KNF UN860FTI EX Explosion Proof Motor Vacuum Pump, PTFE/FFKM; 2.1 cfm/27.6"Hg-20psi

Representative image only.

KNF UN860FTI EX Explosion Proof Motor Vacuum Pump, PTFE/FFKM; 2.1 cfm/27.6"Hg-20psi

KNF | Mfr: 077389/077618| Item: 0705764

Oversized, permanently lubricated bearings for long-term operation

Call For Price

Call For Price

Buy It Together

Current Item

KNF UN860FTI EX Explosion Proof Motor Vacuum Pump, PTFE/FFKM; 2.1 cfm/27.6"Hg-20psi
KNF UN860FTI EX Explosion Proof Motor Vacuum Pump, PTFE/FFKM; 2.1 cfm/27.6"Hg-20psi
0705764

Call For Price
+ =
Total Price 1225000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

1 Year Warranty
Specifications
  • Head ConfigurationSingle Head
  • Wetted PartsPTFE, FFKM
  • Free-Air Capacity (CFM)2.1
  • Free-Air Capacity (LPM)60
  • Free-Air Capacity (LPH)3600
  • Max Flow Rate (CFM)2.1
  • Max Flow Rate (Liters/min)60
  • Max Vacuum (in Hg)27.6
  • Max Vacuum (mbar)78.6
  • Max Vacuum (mm Hg)59
  • Max Vacuum (torr)59
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)6.6/3.3
  • Power (Hz)60
  • Height (in)9.125
  • Width (in)7.375
  • Length (in)19
  • Height (cm)23.18
  • Width (cm)18.73
  • Length (cm)48.26
  • Height (mm)231.8
  • Width (mm)187.3
  • Length (mm)482.6
  • DescriptionExplosion Proof Motor Vacuum Pump, PTFE/FFKM; 2.1 cfm/27.6"Hg-20psi
  • Warranty1 Year
Reviews
Q&A