KNF UN035.1.2STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 2.0 cfm/27"Hg-60psi

Representative image only.

KNF UN035.1.2STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 2.0 cfm/27"Hg-60psi

KNF | Mfr: 072951/075823| Item: 0705776

Oversized, permanently lubricated bearings for long-term operation

Call For Price

Call For Price

Buy It Together

Current Item

KNF UN035.1.2STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 2.0 cfm/27"Hg-60psi
KNF UN035.1.2STI EX Explosion Proof Motor Vacuum Pump, 316SS/PTFE; 2.0 cfm/27"Hg-60psi
0705776

Call For Price
+ =
Total Price 1225000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

1 Year Warranty
Specifications
  • Wetted Parts316 Stainless Steel, PTFE
  • Free-Air Capacity (CFM)2
  • Free-Air Capacity (LPM)56.1
  • Free-Air Capacity (LPH)3366
  • Max Flow Rate (CFM)2.3
  • Max Flow Rate (Liters/min)56.1
  • Max Vacuum (in Hg)27
  • Max Vacuum (mbar)100
  • Max Vacuum (mm Hg)74.2
  • Max Vacuum (torr)74.2
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)6.0
  • Power (Hz)50/60
  • Height (in)8.625
  • Width (in)10.25
  • Length (in)19.625
  • Height (cm)21.91
  • Width (cm)26.04
  • Length (cm)49.85
  • Height (mm)219.1
  • Width (mm)260.4
  • Length (mm)498.5
  • DescriptionExplosion Proof Motor Vacuum Pump, 316SS/PTFE; 2.0 cfm/27"Hg-60psi
  • Warranty1 Year
Reviews
Q&A