KNF Service Kit for LABOPORT® Diaphragm Vacuum Pumps 78160-00, -05 and 78162-00, -05.
Representative image only.
KNF Service Kit for LABOPORT® Diaphragm Vacuum Pumps 78160-00, -05 and 78162-00, -05.
KNF | Mfr: 072778| Item: 7816250
Service kits to keep your vacuum running at its best!
₹48,100.00INR / Each
₹48,100.00INR / Each
Buy It Together
Current Item
KNF Service Kit for LABOPORT® Diaphragm Vacuum Pumps 78160-00, -05 and 78162-00, -05.
KNF Service Kit for LABOPORT® Diaphragm Vacuum Pumps 78160-00, -05 and 78162-00, -05.
7816250
₹48,100.00INR / Each
+ =
Total Price 745100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These service kits help keep your diaphragm vacuum pumps running smoothly. The kits only service one pump head. Two service kits are required for two-stage pumps.
The kit includes one PTFE coated diaphragm and two FFPM valves, plates/seal rings for models.
1 Year Warranty
Specifications
  • Compatible WithVacuum Pumps 78160-00, -05 and 78162-00, -05
  • DescriptionService kit for portable pump 78160-00, -05 and 78162-00, -05