KNF K035-0STA Service Kit for Portable and Explosion-Proof Laboratory Pumps w/ PTFE & SS
Representative image only.
KNF K035-0STA Service Kit for Portable and Explosion-Proof Laboratory Pumps w/ PTFE & SS
KNF | Mfr: 071734| Item: 0705619
Service kits to keep your vacuum running at its best!
₹33,000.00INR / Each
₹33,000.00INR / Each
Buy It Together
Current Item
KNF K035-0STA Service Kit for Portable and Explosion-Proof Laboratory Pumps w/ PTFE & SS
KNF K035-0STA Service Kit for Portable and Explosion-Proof Laboratory Pumps w/ PTFE & SS
0705619
₹33,000.00INR / Each
+ =
Total Price 730000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These service kits help keep your diaphragm vacuum pumps running smoothly. The kits only service one pump head. Two service kits are required for two-stage pumps.
Use with 07056-30, -31, -35, -36 & -50.
Specifications
  • Compatible WithVacuum pumps 07056-30, -31, -35, -36, and -50
  • DescriptionService Kit for Portable and Explosion-Proof Laboratory Pumps w/ PTFE & SS