KleenGuard 3052 KLEENGUARD* G80 NEOPRENE GLOVE
Representative image only.
KleenGuard 3052 KLEENGUARD* G80 NEOPRENE GLOVE
KleenGuard | Mfr: 03052| Item: 3346603
Choose from two chemical resistant materials to fit a wide range of applications
₹31,200.00INR / Case of 144
₹31,200.00INR / Case of 144
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Kleenguard G80 gloves feature cotton flock lining making the gloves easier to put on and remove. Lining also absorbs perspiration making the glove more comfortable. The exclusive glove shape is optimized for the best possible fit on more than forty different dimensions.||Choose the purple nitrile for protection from fuels, grease and solvents.||The neoprene latex blend is resistant to acids, detergents, and caustics.|
Specifications
  • MaterialNeoprene
  • SterileNo
  • Description3052 KLEENGUARD* G80 NEOPRENE GLOVE