KleenGuard 3033 KLEENGUARD* A10 XL W/OUT HOOD
Representative image only.
KleenGuard 3033 KLEENGUARD* A10 XL W/OUT HOOD
KleenGuard | Mfr: 03033| Item: 3343302
Choose the breathable coverall that best meets the requirement of your application
₹7,000.00INR / Case
₹7,000.00INR / Case
Product Description
Key Features
  • Simplifies aseptic gowning
  • Save time and reduce the risk of contamination
More About this Item
Coveralls protect you from dry particulates and light liquid sprays. KleenGuard A5, A10 coveralls are made of a patented microforce™ barrier three-layer fabric which includes a layer of microporous film. This design allows heat and sweat vapours escape so workers to stay with the task at hand with less fatigue and heat stress. The concealed front zipper measures at least 28" long. Three-thread overlap stitching improves strength and durability. Coveralls can be easily donned or removed without tearing. Comfortable and generously cut coveralls are 7 1⁄2 times less likely to rip out than those made using the typical ANSI-standard design. All coveralls feature elastic cuffs, wrists, and back for improved fit and comfort. Sleeve is folded with top portion folded and cuffed over the bottom (wrist) portion, exposing the inside to assist in aseptic donning. Innovative Cuff Fold pattern presents the inside of the sleeves as the package is opened, reducing the risk of touching and contaminating the outside of the sleeves. Integrated Thumb Loop keeps sleeve from rolling back and exposing arm or wrist.
Each individually bagged pair of sleeves has a sterilization indicator as a visual indicator that it has undergone gamma sterilization.
A10 Light duty coveralls are constructed from spun bound polypropylene. Coveralls are breathable helping keep workers cool and comfortable in a wide range of work situations.
A5 Sterile Cleanroom Apparel are made from 100% Polypropylene Breathable SMS Fabric & are suitable for ISO Class 5 or higher cleanrooms. They are CE Marked & comes under Category III, PPE class. Manufactured in ISO9001:2000 facility A5 Sterile Cleanroom Apparel are triple bagged, vacuum packaged, includes sterility indicator folded inside out or cuffed for aseptic donning & No Silicone, No Latex, No BHT preservative are present in them.
Catalogue No:DescriptionMaterialSizeColourQty/Cs
33433-01A10 Sterile Cleanroom CoverallPolypropyleneLargeWhite25
33433-02A10 Sterile Cleanroom CoverallPolypropyleneX-largeWhite25
15945-64A5 Sterile Cleanroom CoverallPolypropyleneMediumWhite25
15945-65A5 Sterile Cleanroom CoverallPolypropyleneLargeWhite25
15945-66A5 Sterile Cleanroom CoverallPolypropyleneX-largeWhite25
15945-67A5 Sterile Cleanroom CoverallPolypropyleneXX-largeWhite25
15945-68A5 Sterile Cleanroom BootsPolypropylene-White100
15945-69A5 Sterile Integrated Hood and MaskPolypropylene-White7
Specifications
  • DescriptionKLEENGUARD* A10 XL W/OUT HOOD