KleenGuard 08582 Safety Glasses, Clear Lens
Representative image only.
KleenGuard 08582 Safety Glasses, Clear Lens
KleenGuard | Mfr: 08582| Item: 1594053
Polycarbonate lens provide a light-weight, comfortable fit
₹57,500.00INR / Case of 1
₹57,500.00INR / Case of 1
Product Description
Key Features
  • Available in a variety of options - V10, V20, V30 and V40 safety glasses
  • Impact resistant
  • Offer 99.9% UVA/UVB protection
More About this Item
KLEENGUARD safety glasses are impact resistant and provide 99.9% UVA/UVB protection.
All glasses meet ANSI Z87.1+ and EN 166 standards.
KLEENGUARD V10 Glasses feature lightweight frames and a choice of clear, amber, or clear OTG (over the glasses) lens.
KLEENGUARD V20 Glasses have wraparound lenses for better eye coverage and padded temples for added comfort. Choose from smoke, anti-fog clear, and indoor/outdoor lenses.
KLEENGUARD V30 Glasses feature wraparound lenses along with padded nose bridge and temples and adjustable lens angle. Choose from amber, anti-fog clear, and indoor/outdoor lenses.
KLEENGUARD V40 Glasses are the ultimate in comfort. Dual-molded rubber temples and cushioned brow bar give a comfortable feel while adjustable soft nose bridge and lens angle allow you to customize the fit. Silver mirror lens is perfect for outdoor environments.
Conformite Europeenne
Specifications
  • Frame ColorClear
  • Lens ColorClear
  • ComplianceANSI Z87.1; EN 166
  • DescriptionSafety Glasses, Clear Lens