Kingscote 506250-R4 Red B Leak Tracer Dye, Solvent-Based, 4 oz
Representative image only.
Kingscote 506250-R4 Red B Leak Tracer Dye, Solvent-Based, 4 oz
Kingscote | Mfr: 506250-R4| Item: 0029504
Provides leak detection and color coding for oils, synthetic oils, fats, waxes, resins and most solvents
₹7,800.00INR / Each
₹7,800.00INR / Each
Product Description
Key Features
  • Liquid, high strength dyes with a wide range of compatability
  • Free from any insoluables
  • 3 month warranty
More About this Item
Dyes diffuse rapidly, producing a vivid, easy-to-see color-- ideal for hydraulics, automotive, color coding, and manufacturing. Dilution factors for coloring light oils is 1 to 64,000; for coloring dark oils is 1 to 12,800; and for leak detection is 1 to 640.
3 Month Warranty
Specifications
  • DescriptionRed B Leak Tracer Dye, Solvent-Based, 4 oz