Keck Sieve Shaker Kit, stainless steel screens, acrylic cylinders, ABS frame
Representative image only.
Keck Sieve Shaker Kit, stainless steel screens, acrylic cylinders, ABS frame
Keck | Mfr: 81450001| Item: 5997810
Lightweight, compact design – easy to take into the field
₹70,500.00INR / Each
₹70,500.00INR / Each
Product Description
Key Features
  • Kit includes 20 interchangeable screens in a wide range of mesh sizes
  • Provides clear indication of sample
  • A complete kit with proven, reliable results
  • 1 year warranty
More About this Item
This complete mechanical sieve kit provides reliable particle size analysis. Kit includes 20 stainless steel mesh screens and five interlocking cylinders. Shakers are easy to use; place the sample in the top cylinder and shake to distribute particles — the volumetric percentages are indicated on the shaker frame. The clear acrylic cylinders make it easy to see sample and accurately read volume. Each cylinder measures 1.85”L x 2” ID; ABS frame measures 14-1/2”L x 4” ID.
Sieve Sizes Included:
0.187” opening (No. 4 U.S. standard)
0.132” opening (No. 6 U.S. standard)
0.090” opening (No. 8 U.S. standard)
0.072” opening (No. 10 U.S. standard)
0.060” opening (No. 12 U.S. standard)
0.051” opening (No. 14 U.S. standard)
0.046” opening (No. 16 U.S. standard)
0.040” opening (No. 18 U.S. standard)
0.030” opening (No. 20 U.S. standard)
0.026” opening (No. 25 U.S. standard)
0.023” opening (No. 30 U.S. standard)
0.020” opening (No. 35 U.S. standard)
0.015” opening (No. 40 U.S. standard)
0.009” opening (No. 60 U.S. standard)
0.0055” opening (No. 100 U.S. standard)
0.0046” opening (No. 120 U.S. standard)
0.0041” opening (No. 140 U.S. standard)
0.0029” opening (No. 200 U.S. standard)
0.0024” opening (No. 230 U.S. standard)
0.0021” opening (No. 270 U.S. standard)
1 Year Warranty
Specifications
  • MaterialABS frame, acrylic cylinders
  • DescriptionSieve Shaker Kit, stainless steel screens, acrylic cylinders, ABS frame