Justrite UI25S Steel Safety Can for Flammables, Type I, Flame Arrester, 2.5 Gal

Representative image only.

Justrite UI25S Steel Safety Can for Flammables, Type I, Flame Arrester, 2.5 Gal

Justrite | Mfr: UI25S| Item: 8179142

Safely contain hazardous liquids and control vapors

₹19,000.00INR / Each

₹19,000.00INR / Each

Buy It Together

Current Item

Justrite UI25S Steel Safety Can for Flammables, Type I, Flame Arrester, 2.5 Gal
Justrite UI25S Steel Safety Can for Flammables, Type I, Flame Arrester, 2.5 Gal
8179142

₹19,000.00INR / Each
+ =
Total Price 25000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
  • 10 year warranty

More About this Item

All cans include a 3 1/2” built-in, stainless-steel flame arrester to prevent flashback ignition. Self-closing, spring loaded lid automatically vents at 3-5 psi (0.2 and 0.35 bar) to fight rupture or explosion. Reinforcing ribs resist denting or implosion. Steel construction with powder coat paint resists corrosion and improves wear and tear. A large ID zone allows you to label content of the can to avoid misuse

Type I safety cans feature one opening that serves for both filling and pouring. Leakproof lid protects vapors from escaping and guards against spills.

FM Approved10 Year Warranty
Specifications
  • MaterialSteel
  • Capacity (Gal)2.5
  • Capacity (Liters)9.46
  • Exterior Height (in)11.5
  • Exterior Diameter (in)11.75
  • Exterior Height (cm)29.21
  • Exterior Diameter (cm)29.85
  • ComplianceNFPA, OSHA
  • ApprovalFM, UL, cUL
  • DescriptionSafety Can for Flammables, Type I, 2.5 gallon
  • Warranty10 Years
Reviews
Q&A