Justrite Quick-Disconnect Disposal Can, Stainless Steel Fittings; 2 Gal

Representative image only.

Justrite Quick-Disconnect Disposal Can, Stainless Steel Fittings; 2 Gal

Justrite | Mfr: TF12752| Item: 1203072

Remove can and empty waste without disconnecting your inlet tubing

₹252,000.00INR / Each

₹252,000.00INR / Each

Buy It Together

Current Item

Justrite Quick-Disconnect Disposal Can, Stainless Steel Fittings; 2 Gal
Justrite Quick-Disconnect Disposal Can, Stainless Steel Fittings; 2 Gal
1203072

₹252,000.00INR / Each
+ =
Total Price 303500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Stainless steel fittings for 3/8" tubing.

Key Features

  • Safe method to stage and draw reagents
  • Quick disconnects allow simple attachment of one inlet and one vent
  • Eliminate safety concerns and accidents associated with glass bottles
  • FM approved
  • EPA complaint

More About this Item

Quick-disconnect safety cans provide a safe hands-free way to drain waste from your HPLC application. Prevent the escape of ignitable and toxic vapors. Protect the liquid and vapors inside from external heat sources. The quick disconnect mechanisms are valved open when connected and valved closed when disconnected for spill free changeout of safety cans without disconnecting the tubing.

Coalescing carbon filter attaches to one of the fittings to filter VOCs. Durable polyethylene is translucent allowing you quick visual inspection of liquid level. Pouring spout includes stainless steel flame arrester to prevent flashback ignition. Dual fittings eliminate back pressure and provide controlled handling of vapors. Container automatically vents at 3-5 psig.

An optional spill basin (86394-45) captures any leakage and provides additional protection against accidental pumping.

Specifications
  • Capacity (Gal)2
  • Capacity (Liters)8
  • Exterior Height (in)14.75
  • Exterior Diameter (in)12
  • Exterior Height (cm)37.5
  • Exterior Diameter (cm)30.5
  • ApprovalFM
  • DescriptionQuick-Disconnect Disposal Can; 2 Gal
Reviews
Q&A