Julabo Thermal P60 5L, clear Presto fluid
Representative image only.
Julabo Thermal P60 5L, clear Presto fluid
Julabo | Mfr: 8891413| Item: 1214971
Quality accessories for Julabo Recirculators and Presto systems
₹373,000.00INR / Each
₹373,000.00INR / Each
Product Description
Key Features
  • Temperature range is -60 to 250 degC
  • Flash point at 443 deg C
  • 2 year warranty
More About this Item
This clear, odorless liquid covers a wide rang of temperatures. Low viscosity, and a long shelf life make it a good choice for the Presto baths.
Two year warranty
Specifications
  • Description5L, clear Presto fluid