Julabo Lockable Bath Lid for 600F, 601F, 1000F, 1000FW, and BC12 Refrigerated/Heating Circulators
Representative image only.
Julabo Lockable Bath Lid for 600F, 601F, 1000F, 1000FW, and BC12 Refrigerated/Heating Circulators
Julabo | Mfr: 9970243| Item: 1215374
For use with the MAGIO™ range of refrigerated, heated, and immersion circulators/baths
₹639,000.00INR / Each
₹639,000.00INR / Each
Product Description
Key Features
  • Optimize performance of your refrigerated/heating circulators with these accessories
  • Minimize oil vapour and odour formation at high temperatures
  • Bath cover is lockable
  • 2 Years Part and Labor warranty
More About this Item
Choose from accessories compatible with the MAGIO™ range of circulators include an electronic module to allow for analog connections to the pump, a condensation trap with lid, and lockable lid. Both bath lid accessories increase energy efficiency by preventing steam leakage and the associated fluid loss.
2 Year Warranty
Specifications
  • Compatible WithJulabo MAGIO, CORIO CP and CD, DYNEO DD models BC12, 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
  • DescriptionLockable Bath Lid for 600F, 601F, 1000F, 1000FW, and BC12 Refrigerated/Heating Circulators
  • Warranty2 Years Part and Labor warranty