Julabo CORIO™ Test Tube Rack, SS, for 10 Falcon Tubes; 50 mL

Representative image only.

Julabo CORIO™ Test Tube Rack, SS, for 10 Falcon Tubes; 50 mL

Julabo | Mfr: 9970323| Item: 1214728

Quality accessories for Julabo Recirculators and Presto systems

₹71,300.00INR / Each

₹71,300.00INR / Each
Product Description

Key Features

  • Good for use up to +150°C for CORIO™ CD-B5 and -BT5 open bath tanks
  • 2 year warranty
Two year warranty
Specifications
  • Bath TypeBath/Circulator Accessory
  • DescriptionTest Tube Rack, Stainless Steel, for 10 Falcon Tubes; 50 mL
Reviews
Q&A