Julabo CORIO™ Test Tube Rack, Plastic, for 21 Tubes; 30 mm

Representative image only.

Julabo CORIO™ Test Tube Rack, Plastic, for 21 Tubes; 30 mm

Julabo | Mfr: 9970303| Item: 1214732

Quality accessories for Julabo Recirculators and Presto systems

₹41,600.00INR / Each

₹41,600.00INR / Each
Product Description

Key Features

  • Good for use up to +100°C for B13, B17, B19, B27, B33, BT19, and BT27 open bath tanks
  • 2 year warranty
Two year warranty
Specifications
  • Bath TypeBath/Circulator Accessory
  • DescriptionTest Tube Rack, Plastic, for 21 Tubes; 30 mm
Reviews
Q&A