Julabo Condensation Trap with Bath Lid for 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
Representative image only.
Julabo Condensation Trap with Bath Lid for 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
Julabo | Mfr: 9970700| Item: 1215311
For use with the MAGIO™ range of refrigerated, heated, and immersion circulators/baths
₹802,000.00INR / Each
₹802,000.00INR / Each
Product Description
Includes
condensation trap and bath lid.
Key Features
  • Optimize performance of your refrigerated/heating circulators with these accessories
  • Suitable for continuous operation at temperatures < -10°C
  • 2 Years Product / 2 Year Parts and Labor Warranty
More About this Item
Choose from accessories compatible with the MAGIO™ range of circulators include an electronic module to allow for analog connections to the pump, a condensation trap with lid, and lockable lid. Both bath lid accessories increase energy efficiency by preventing steam leakage and the associated fluid loss.
The condensation trap reduces the dilution of alcoholic and oil-based bath fluids. The trap contains a cooling element, which causes the humidity of passing air to condense or freeze depending on temperature.
Specifications
  • Compatible WithJulabo MAGIO MS, CORIO CP and CD, DYNEO DD models 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
  • DescriptionCondensation Trap with Bath Lid for 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
  • Warranty2 Years Product / 2 Year Parts and Labor