Julabo Clamp for 500 mL Erlenmeyer flask; 8 flasks held in bath

Representative image only.

Julabo Clamp for 500 mL Erlenmeyer flask; 8 flasks held in bath

Julabo | Mfr: 8 970 608| Item: 1205055

Call For Price

for an additional
₹68,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Julabo Clamp for 500 mL Erlenmeyer flask; 8 flasks held in bath
Julabo Clamp for 500 mL Erlenmeyer flask; 8 flasks held in bath
1205055

Call For Price
+
InnoCal NIST-Traceable Calibration; Temperature Baths
1700113

₹68,800.00INR / Each
=
Total Price 68800.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty
1 Year Warranty
Specifications
  • Reservoir Capacity (L)0.5
  • DescriptionClamp for 500 mL Erlenmeyer flask; 8 flasks held in bath
  • Warranty1 year
Reviews
Q&A