Jenway 970 231B Dissolved Oxygen Probe for 970 Meters
Representative image only.
Jenway 970 231B Dissolved Oxygen Probe for 970 Meters
Jenway | Item: 9996491
Jenway has an extensive range of dissolved oxygen probes suitable for portable or bench dissolved oxygen meters
₹70,800.00INR / Each
₹70,800.00INR / Each
Product Description
Key Features
  • The probes for the dissolved oxygen meters have all been updated in order to unify the range
More About this Item
The dissolved oxygen probes have been redesigned so that they are all the same diameter. As a result of this redesign, the new probes have a narrower diameter than the old probes. The new probes are easily identified with a B after the probe part number on the probe or meter packaging.
To compliment the new probe design the dissolved oxygen membranes have also been redesigned. The new membranes will not fit on old probes and old membranes will not fit on new probes.
Specifications
  • Accessory TypeDissolved oxygen probe
  • DescriptionDissolved Oxygen Probe for 970 Meters