Jenway 924 076S pH Electrode Fill Solution, 4M KCL, Ag/AgCl; 150 mL

Representative image only.

Jenway 924 076S pH Electrode Fill Solution, 4M KCL, Ag/AgCl; 150 mL

Jenway | Item: 7900085

All your pH measurement needs in one place

₹9,500.00INR / Each

₹9,500.00INR / Each

Buy It Together

Current Item

Jenway 924 076S pH Electrode Fill Solution, 4M KCL, Ag/AgCl; 150 mL
Jenway 924 076S pH Electrode Fill Solution, 4M KCL, Ag/AgCl; 150 mL
7900085

₹9,500.00INR / Each
+ + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 33600.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles
  • Fill solution for electrode 79000-84 (924 076). Probe sold separately.

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications
  • Accessory TypepH Electrode Fill Solution
  • Range4M KCL, Ag/AgCl; 150 mL
  • DescriptionpH Electrode Fill Solution, 4M KCL, Ag/AgCl; 150 mL
Reviews
Q&A