Jenway 025 197 pH Electrode Partial Gel, 4M KCl, AgCl; 150 mL

Representative image only.

Jenway 025 197 pH Electrode Partial Gel, 4M KCl, AgCl; 150 mL

Jenway | Item: 7900039

All your pH measurement needs in one place

₹4,200.00INR / Each

₹4,200.00INR / Each

Buy It Together

Current Item

Jenway 025 197 pH Electrode Partial Gel, 4M KCl, AgCl; 150 mL
Jenway 025 197 pH Electrode Partial Gel, 4M KCl, AgCl; 150 mL
7900039

₹4,200.00INR / Each
+ + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 28300.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications
  • Accessory TypepH Electrode Partial Gel
  • Range4M KCl, AgCl; 150 mL
  • DescriptionpH Electrode Partial Gel, 4M KCl, AgCl; 150 mL
Reviews
Q&A