Jenway 025 192 pH Electrode Storage Solution; 100 mL

Representative image only.

Jenway 025 192 pH Electrode Storage Solution; 100 mL

Jenway | Item: 9995960

All your pH measurement needs in one place

₹6,400.00INR / Each

₹6,400.00INR / Each

Buy It Together

Current Item

Jenway 025 192 pH Electrode Storage Solution; 100 mL
Jenway 025 192 pH Electrode Storage Solution; 100 mL
9995960

₹6,400.00INR / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+ + =
Total Price 30500.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Jenway offers a range of pH buffers either as single use sachets or longer lasting 500 mL bottles

More About this Item

To care for your Jenway pH electrodes there is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications
  • Accessory TypepH Electrode Storage Solution
  • Range100 mL
  • DescriptionpH Electrode Storage Solution; 100 mL
Reviews
Q&A