Jenway 025 165 1382 ppm TDS Calibration Standard, 500 mL

Representative image only.

Jenway 025 165 1382 ppm TDS Calibration Standard, 500 mL

Jenway | Item: 7900035

Solutions for calibration of Jenway conductivity meters

₹4,900.00INR / Each

₹4,900.00INR / Each
Product Description
Specifications
  • Conductivity Value1382 ppm
  • Volume500 mL
  • Use withJenway conductivity meters
  • Description1382 ppm TDS Calibration Standard, 500 mL
Reviews
Q&A