Jackson Safety® Maxview™ Clear Polycarbonate Face Shield for Slotted Hard-Hats
Representative image only.
Jackson Safety® Maxview™ Clear Polycarbonate Face Shield for Slotted Hard-Hats
| Mfr: SEL14202| Item: 8647804
Offers maximum protection against splash and flying debris
₹9,500.00INR / Each
₹9,500.00INR / Each
Product Description
Key Features
  • Large polycarbonate window offers superior optics
  • Side and chin guard protects against splash and flying debris
  • Extended crown safeguards against overhead hazards
  • Meets ANSI Z87.1 and CSA Z94.3 standards
More About this Item
The Maxview face shields offer maximum face protection coverage with a high impact polycarbonate window. The oversized window provides you with a clear panoramic view. A built-in side and chin guard along with an extended top of head coverage protects you against splash and flying debris as well as overhead hazards.
Choose from anti-fog coating and slotted hard hat mounting face shields.
Specifications
  • Frame ColorBlack
  • Lens ColorClear
  • Lens CoatingNone
  • ComplianceANSI Z81.1 high impact, CSA Z94.3
  • Width (in)9 1/16
  • Height (in)13 3/8
  • Depth (in)1/25
  • DescriptionClear Polycarbonate Face Shield for Slotted Hard-Hats