ITS 481396 Water Quality Test Kit , Arsenic, Standard Range , 100/Pk
Representative image only.
ITS 481396 Water Quality Test Kit , Arsenic, Standard Range , 100/Pk
ITS | Mfr: 481396| Item: 9953132
Measure arsenic levels in drinking water in only 12 minutes
₹51,800.00INR / Pkg of 100
₹51,800.00INR / Pkg of 100
Product Description
Key Features
  • Accurate, reliable results down to 2 ppb
  • Included in the EPA's Environmental Technology Verification program
More About this Item
Quickly and safely analyze samples. Only 2 minutes required for preparation, and only 10 minutes for incubation with this easy-to-use test kit. Reagents are in powder form and were carefully selected to minimize hazards normally associated with conventional arsenic testing kits.
Specifications
  • TestArsenic, standard range
  • Measurement Points0, 5, 10, 20, 40, 60, 100, 200, 300, and 500 ppb
  • Tests Per Kit100
  • DescriptionWater Quality Test Kit , Arsenic, Standard Range , 100/Pk