ITS 481297 Water Quality Test Kit , Arsenic, Low Range , 50/Pk
Representative image only.
ITS 481297 Water Quality Test Kit , Arsenic, Low Range , 50/Pk
ITS | Mfr: 481297| Item: 9953134
Measure arsenic levels in drinking water in only 12 minutes
₹58,200.00INR / Pkg of 50
₹58,200.00INR / Pkg of 50
Product Description
Key Features
  • Accurate, reliable results down to 2 ppb
  • Included in the EPA's Environmental Technology Verification program
More About this Item
Quickly and safely analyze samples. Only 2 minutes required for preparation, and only 10 minutes for incubation with this easy-to-use test kit. Reagents are in powder form and were carefully selected to minimize hazards normally associated with conventional arsenic testing kits.
Specifications
  • TestArsenic, low range
  • Measurement Points0, 2, 4, 6, 8, 10, 15, 20, 30, 40, 60, 100, 160, and 240 ppb
  • Tests Per Kit50
  • DescriptionWater Quality Test Kit , Arsenic, Low Range , 50/Pk