ITS 481026 Water Quality Test Strips, Free Chlorine, 0 to 5.0 ppm, 50/Bottle
Representative image only.
ITS 481026 Water Quality Test Strips, Free Chlorine, 0 to 5.0 ppm, 50/Bottle
ITS | Mfr: 481026| Item: 9953120
Obtain test results within 90 seconds
₹6,700.00INR / Each
₹6,700.00INR / Each
Product Description
Key Features
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
More About this Item
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
Specifications
  • TestChlorine, free
  • Measurement Points0, 0.05, 0.1, 0.2, 0.4, 0.5, 0.8, 1.0, 2.0, 5.0 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionWater Quality Test Strips, Free Chlorine, 0 to 5.0 ppm, 50/Bottle