ITS 480014 Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottle
Representative image only.
ITS 480014 Water Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottle
ITS | Mfr: 480014| Item: 9953130
Obtain test results within 90 seconds
₹5,700.00INR / Each
₹5,700.00INR / Each
Product Description
Key Features
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
More About this Item
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
Specifications
  • TestPeroxide
  • Measurement Points0, 0.5, 2.0, 5.0, 10, 25, 50, 100 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionWater Quality Test Strips, peroxide, 0 to 100 ppm, 50/bottle