ITS 480010 Water Quality Test Strips, total chlorine, 0 to 10.0 ppm, 50/bottle
Representative image only.
ITS 480010 Water Quality Test Strips, total chlorine, 0 to 10.0 ppm, 50/bottle
ITS | Mfr: 480010| Item: 9953122
Obtain test results within 90 seconds
₹7,700.00INR / Each
₹7,700.00INR / Each
Product Description
Key Features
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
More About this Item
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
Specifications
  • TestChlorine, total
  • Measurement Points0, 0.05, 0.075, 0.1, 0.125, 0.15, 0.175, 0.2, 0.3, 0.5, 0.8, 4.0, 10.0 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionWater Quality Test Strips, total chlorine, 0 to 10.0 ppm, 50/bottle