ITS 480008 Water Quality Test Strips, total hardness, 0 to 425 ppm, 50/bottle
Representative image only.
ITS 480008 Water Quality Test Strips, total hardness, 0 to 425 ppm, 50/bottle
ITS | Mfr: 480008| Item: 9953103
Obtain test results within 90 seconds
₹3,200.00INR / Each
₹3,200.00INR / Each
Product Description
Key Features
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
  • 1 month warranty
More About this Item
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
1 Month Warranty
Specifications
  • TestHardness, total
  • Measurement Points0 to 425 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionWater Quality Test Strips, total hardness, 0 to 425 ppm, 50/bottle
  • Warranty1 month