ITS 480005 Water Quality Test Strips, PH/Total Alkalinity (Ta), 6-9 PH/0-360 Ta, 50/Bottle
Representative image only.
ITS 480005 Water Quality Test Strips, PH/Total Alkalinity (Ta), 6-9 PH/0-360 Ta, 50/Bottle
ITS | Mfr: 480005| Item: 9953128
Obtain test results within 90 seconds
₹5,400.00INR / Each
₹5,400.00INR / Each
Product Description
Key Features
  • Test strips feature bright easy-to-read colors
  • No hazardous reagents
More About this Item
These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.
Specifications
  • TestpH and total alkalinity
  • Measurement Points6 to 9 pH; 0, 80, 120, 180, 240, 360 TA
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionWater Quality Test Strips, PH/Total Alkalinity (Ta), 6-9 PH/0-360 Ta, 50/Bottle