International Products Corp Surface-Cleanse/930 Liquid Concentrate; 20 L
Representative image only.
International Products Corp Surface-Cleanse/930 Liquid Concentrate; 20 L
International Products Corp | Mfr: S-2020| Item: 1810330
Easily clean glass, metal, plastic, rubber, and other hard materials
₹122,000.00INR / Each
₹122,000.00INR / Each
Product Description
Key Features
  • Leaves no charged residue—perfect for cleaning electrical components!
  • 1 year warranty
More About this Item
Safely wash away grease, petroleum, fingerprints, oils, scale, oxides, and metallic residues from glass, aluminum, nickel, lead, zinc, ceramic, and plastic without surface damages typically caused by acidic or alkaline cleaners. Use to clean filter membranes and other delicate items. Non-flammable and non-hazardous under U.S. federal regulations for shipping, storage, and disposal. Meets USDA guidelines for incidental contact with food processing systems. Contains no CFCs, phosphates, silicates, borates, halogens, or phenols.
Surface-Cleanse/930 is a gentle, non-ionic cleaner designed to deliver powerful cleaning action. It contains a mixture of nonionic sufacatants that is safe for use on aluminum, zinc and other delicate metals and the cleaner's low sodium content makes it ideal for use on electronic components. Its neutral pH eliminates surface damage that may be caused by an acid or alkaline cleaner.
Target soils include adhesive, oil, flux and ink. Solution is safe for workers and the environment - contains no phosphates, CFCs, ODCs, phenols or borates. Designed for use in CIP, ultrasonic and immersion applications. A 1% to 2% solution is effective in most appilcations. Typical pH: 9.5. NSF registered for USDA A1 use.
1 Year Warranty
Specifications
  • FormatLiquid
  • Volume (gallons)5
  • Volume (Liters)20
  • Dilution1:200, 7 pH
  • DescriptionSurface-Cleanse/930 Liquid Concentrate, 20 L