International Products Corp M-0801-12 Micro A07 Citric Acid Liquid Cleaner, 1 Liter
Representative image only.
International Products Corp M-0801-12 Micro A07 Citric Acid Liquid Cleaner, 1 Liter
International Products Corp | Mfr: M-0801-12| Item: 1810400
Environmentally friendly – Phosphate Free
₹9,500.00INR / Each
₹9,500.00INR / Each
Product Description
Key Features
  • Free Rinsing
More About this Item
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)1
  • Dilution1:100 to 2:100, 2.5 pH
  • DescriptionMicro A07 Citric Acid Liquid Cleaner, 1 Liter