International Products Corp M-0801-12 Micro A07 Citric Acid Cleaner, 12/cs
Representative image only.
International Products Corp M-0801-12 Micro A07 Citric Acid Cleaner, 12/cs
International Products Corp | Mfr: M-0801-12| Item: 1810401
Environmentally friendly – Phosphate Free
₹75,300.00INR / Case of 1
₹75,300.00INR / Case of 1
Product Description
Key Features
  • Free Rinsing
  • Case of 12 1L bottles
More About this Item
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.
Specifications
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)1
  • Dilution1:100 to 2:100, 2.5 pH
  • DescriptionMicro A07 Citric Acid Cleaner case of 12 1L bottles