InterLab PolySeed® NX Microbial CBOD Capsules; 50/Pk
Representative image only.
InterLab PolySeed® NX Microbial CBOD Capsules; 50/Pk
| Mfr: D1255| Item: 5320034
Get fresh consistent seed each time
₹59,000.00INR / Pkg of 50
₹59,000.00INR / Pkg of 50
Product Description
Includes
50 capsules per bottle.
Key Features
  • One easy-to-use capsule performs approximately 100 tests
  • 3 month warranty
More About this Item
Polyseed NX capsules contain a chemical additive to inhibit nitrogenous oxygen demand when performing CBOD.
3 Month Warranty
Specifications
  • Buffer Kit Values50
  • DescriptionMicrobial CBOD Capsules; 50/Pk