InterLab PolySeed® Microbial BOD Capsules; 50/Pk
Representative image only.
InterLab PolySeed® Microbial BOD Capsules; 50/Pk
| Mfr: D1250| Item: 5320033
Get fresh consistent seed each time
₹42,000.00INR / Pkg of 50
₹42,000.00INR / Pkg of 50
Product Description
Includes
50 capsules per bottle.
Key Features
  • One easy-to-use capsule performs approximately 100 tests
  • 3 month warranty
More About this Item
Polyseed capsules contain a special blend of microbial cultures that provide a uniform standard for BOD analysis.
3 Month Warranty
Specifications
  • Buffer Kit Values50
  • DescriptionMicrobial BOD Capsules; 50/Pk