InnoCal NIST-Traceable Calibration, Resistance/Decade Box
Representative image only.
InnoCal NIST-Traceable Calibration, Resistance/Decade Box
InnoCal | Item: 1710135
NIST-traceable calibrations for resistance testers
₹42,700.00INR / Each
₹42,700.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration, Resistance/Decade Box
InnoCal NIST-Traceable Calibration, Resistance/Decade Box
1710135
₹42,700.00INR / Each
+ + + =
Total Price 85400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a resistance/decade box
  • 1 year warranty
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
1 Year Warranty
Specifications
  • Calibration TypeElectrical
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsCalibrated per manufacturer's procedure
  • DescriptionNIST-Traceable Calibration; Resistance Decade Box