InnoCal NIST-Traceable Calibration, Pressure Module
Representative image only.
InnoCal NIST-Traceable Calibration, Pressure Module
InnoCal | Item: 1709026
NIST-traceable calibrations for pressure modules
₹37,100.00INR / Each
₹37,100.00INR / Each
Product Description
Includes
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a pressure module
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Note: Price per module.
NIST Traceable Calibration
Specifications
  • Calibration TypePressure
  • Probe Only CalibrationYes
  • Number Of Calibration Test Points10 Points Ascending and 10 Points Descending
  • Calibration Test PointsAcross the range of the instrument
  • Calibration Test Range0 to 20,000 PSI
  • DescriptionNIST-Traceable Calibration; Pressure Modules