InnoCal NIST-Traceable Calibration, Multimeter with IR Thermometer
Representative image only.
InnoCal NIST-Traceable Calibration, Multimeter with IR Thermometer
InnoCal | Item: 1709008
NIST-traceable calibrations for electrical meters
₹21,600.00INR / Each
₹21,600.00INR / Each
Product Description
Includes
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated.
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a multimeter with IR thermometer
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Calibrations on handheld multimeters with infrared functions are performed using high-accuracy multi-function calibrators and standard-grade reference multimeters per manufacturer’s recommended procedures. The infrared function of the meter is calibrated using radiometrically-calibrated blackbody sources. Electrical measurements can be taken up to 40,000 Volts, 1000 Amps, 10 Tera-Ohms, 110 Mega-Farads, and 1.1 Giga-Hertz and infrared temperatures can be taken between -15 and 500 deg C.
NIST Traceable Calibration
Specifications
  • Calibration TypeElectrical
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsCalibrated per manufacturer's procedure
  • DescriptionNIST-Traceable Calibration; Multimeter with IR Thermometer