InnoCal NIST-Traceable Calibration; Dry-Well Calibrator, Single
Representative image only.
InnoCal NIST-Traceable Calibration; Dry-Well Calibrator, Single
InnoCal | Item: 1700216
From single dry-well to infrared calibrators, let InnoCal be your source for NIST-traceable certification.
₹54,000.00INR / Each
₹54,000.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
Specifications
  • Calibration TypeTemperature
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 Points
  • Calibration Test PointsAcross the range of instrument
  • Calibration Test Range-80 to 650C
  • DescriptionNIST-Traceable Calibration; Single Dry-Well Calibrator