InnoCal NIST-Traceable Calibration; Carbon Dioxide Meter
Representative image only.
InnoCal NIST-Traceable Calibration; Carbon Dioxide Meter
InnoCal | Item: 1710225
Protect personnel from gas hazards due to inaccurate readings – calibrate your gas detector today!
₹32,100.00INR / Each
₹32,100.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Factory authorized service center for MSA and Sperian gas detectors
More About this Item
InnoCal's specially trained technicians test your gas detector with a known concentration of gas. Calibration drift can occur due to many factors: normal degradation of sensors over time, exposure to extreme temperature and humidity conditions, exposure to high levels of the target gases, and harsh storage and handling conditions such as being dropped or submerged in liquid. Ensure worker safety by having your gas detector calibrated by our factory-trained technicians to verify proper operation.
Specifications
  • Calibration TypeAir and Gas Measurement
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1
  • Calibration Test PointsPer manufacturer's recommendation
  • Calibration Test Range0-5%
  • DescriptionNIST-Traceable calibration for carbon dioxide meters