InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.01 oz to 0.5 lbs, ASTM 0-3; OIML E2, F1, F2
Representative image only.
InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.01 oz to 0.5 lbs, ASTM 0-3; OIML E2, F1, F2
InnoCal | Item: 1710814
ISO/IEC 17025:2005-Accredited Report
₹18,700.00INR / Each
₹18,700.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
More About this Item
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.
Specifications
  • DescriptionNIST Calibration Service for Masses. 0.01 oz to 0.5 Lbs; ASTM 0-3; Oiml E2, F1, F2