InnoCal A2LA ISO17025 Accredited Calibration; Stroboscope
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Stroboscope
InnoCal | Item: 1750040
₹39,900.00INR / Each
₹39,900.00INR / Each
Buy It Together
Current Item
InnoCal A2LA ISO17025 Accredited Calibration; Stroboscope
InnoCal A2LA ISO17025 Accredited Calibration; Stroboscope
1750040
₹39,900.00INR / Each
+
Traceable Noncontact Tachometer with Red LED
9876708
₹21,800.00INR / Each
=
Total Price 61700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • A2LA Accredited Calibration. View our scope of accreditation
  • Calibration service for stroboscopes
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Mechanical
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points10 points
  • Calibration Test PointsThroughout range of instrument
  • Calibration Test Range0 to 120,000 flashes per minute
  • DescriptionA2LA ISO17025 Accredited Calibration; Stroboscope