InnoCal A2LA ISO17025 Accredited Calibration; Dry-Well Calibrator, Dual
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Dry-Well Calibrator, Dual
InnoCal | Item: 1750010
From single dry-well to infrared calibrators, let InnoCal be your source for NIST-traceable certification.
₹84,800.00INR / Each
₹84,800.00INR / Each
Product Description
Key Features
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Temperature
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 Points
  • Calibration Test PointsAcross the range of instrument
  • Calibration Test Range-80 to 650C
  • DescriptionA2LA ISO17025 Accredited Calibration; Dry-Well Calibrator, Dual