InnoCal A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass Set, 5 to 10 kg; ASTM 4-7, Class F
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass Set, 5 to 10 kg; ASTM 4-7, Class F
InnoCal | Item: 1750176
ISO/IEC 17025:2005-Accredited Report
₹84,700.00INR / Each
₹84,700.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • ISO/IEC 17025:2005-Accredited Report
  • A2LA Accredited Calibration. View our scope of accreditation
More About this Item
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.
Specifications
  • DescriptionA2LA ISO17025 Accredited Calibration; Calibration Weight/Mass Set, 5 to 10 kg; ASTM 4-7, Class F