Implen C40 NanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V

Representative image only.

Implen C40 NanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V

Implen | Mfr: C40-Mobile| Item: 8307062

Microvolume measurement with on-the-go analysis

Call For Price

for an additional
₹50,100.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Implen C40 NanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V
Implen C40 NanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V
8307062

Call For Price
+
NIST-Traceable Calibration; Spectrophotometer
1704501

₹50,100.00INR / Each
+ + =
Total Price 1101100.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

USB cable, Ethernet cable, built-in lithium ion battery, and power cord.

Key Features

  • Flexible control and data collection via computer, Mac, smartphone, or tablet
  • Choose a Touch model for a built-in touch screen tablet
  • Slot for a Kensington™ lock provides antitheft security
  • 1 year warranty

More About this Item

For those in need of flexibility, this family of UV/visible spectrophotometers offers reliable protein and DNA measurements for mobile and laboratory applications. Requiring only a minute sample size of 0.3 µL and with a limit of detection down to 0.1 ng/µL (ds DNA) or 0.003 mg/µL (BSA), this spectrophotometer series accommodates any application. Simplified optics with two precisely defined pathlengths provide two fixed and sealed anchor points, prohibiting drift and eliminating the need for recalibration.


Equipped with Wi-Fi, USB A/B, HDMI and LAN interface connections for flexible instrument control and data saving options. Front USB flash drive port offers quick and easy data transfer.


1 Year Warranty
Specifications
  • Source LampXenon
  • Bandwidth< 1.8 nm
  • Min Wavelength (nm)200
  • Max Wavelength (nm)900
  • Wavelength Accuracy±0.75 nm
  • Wavelength Reproducibility±0.2 nm
  • Pathlength (mm)0.67 and 0.07
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)2.6
  • Detector3648 CCD Array
  • Concentration130 ng/µL (ds DNA); 3.7 mg/mL (BSA)
  • Scan Time3.5 to 6 seconds
  • Display TypeTouch screen
  • Width (cm)20
  • Height (cm)12
  • Depth (cm)20
  • Width (in)7 7/8
  • Height (in)4 3/4
  • Depth (in)7 7/8
  • Battery1 rechargeable Lithium-ion (included)
  • Power (VAC)90 to 250
  • Power (Hz)50/60
  • DescriptionNanoPhotometer UV/Vis Spectrophotometer Mobile System, 110/220 V
  • Warranty1 year
Reviews
Q&A