Imada DTXS-40 Digital Cap Torque Tester, 40 lbf-in
Representative image only.
Imada DTXS-40 Digital Cap Torque Tester, 40 lbf-in
Imada | Mfr: DTXS-40| Item: 5984831
₹563,000.00INR / Each
for an additional
₹27,900.00INR / Each
for an additional
₹40,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹563,000.00INR / Each
Buy It Together
Current Item
Imada DTXS-40 Digital Cap Torque Tester, 40 lbf-in
Imada DTXS-40 Digital Cap Torque Tester, 40 lbf-in
5984831
₹563,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Torque Tester/Meter
1708015
₹27,900.00INR / Each
=
Total Price 590900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Programmable high/low setpoints with LED indicators for quick GO/NO GO testing
  • 1000 Data memory for recall and download
  • Accuracy: ±0.5% F.S. ±1 LSD
  • Clamps containers 1/2" to 8" in diameter
  • lbf-in, kgf-cm, and N-cm (selectable)
  • RS-232, Digimatic and analog outputs
  • Clockwise and counterclockwise operation
  • Real time and peak mode (selectable)
  • Tare function
  • AC adapter/charger and carrying case
Two year warranty
Specifications
  • Capacity (lb-in)40
  • Capacity (N-cm)500
  • Resolution (lb-in)0.01
  • Resolution (N-cm)0.1
  • Accuracy±0.5% F.S. ±1 LSD
  • DisplayLED
  • DescriptionDigital Cap Torque Tester, 40 lbf-in