Imada CB-203 Data Acquisition Software RS-232 Cable For DS2
Representative image only.
Imada CB-203 Data Acquisition Software RS-232 Cable For DS2
Imada | Mfr: CB-203| Item: 9395517
Capture key data from your torque testers
₹31,500.00INR / Each
₹31,500.00INR / Each
Buy It Together
Current Item
Imada CB-203 Data Acquisition Software RS-232 Cable For DS2
Imada CB-203 Data Acquisition Software RS-232 Cable For DS2
9395517
₹31,500.00INR / Each
+
Imada OH-1 Optional Handle for Gauges
5985044
₹37,100.00INR / Each
=
Total Price 68600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • 2 year warranty
More About this Item
Gather peak torque or first peak readings as well as a running log of all torque measurements along with a chart of the data and statistical information. All data is entered directly into a standard Excel spreadsheet, allowing you to analyze your torque tester data using all of the powerful tools available in Excel.
RS-232 Cable for DS2 digital force gauges
Two year warranty
Specifications
  • DescriptionData Acquisition Software RS-232 Cable For DS2