Ilmvac Replacment Vacuum Pump Oil
Representative image only.
Ilmvac Replacment Vacuum Pump Oil
Ilmvac | Mfr: 800122| Item: 1594397
Ideal for use with freeze dryers
Call For Price
Call For Price
Product Description
Includes
anti-suction back valve, gas ballast valve, vacuum pump oil, centering ring, clamping ring and motor overload protection, and plug leads (EU, UK and US).
Key Features
  • High water vapor tolerance and reliability
  • 1 year warranty
More About this Item
This two-stage rotary vane pump is designed to withstand large amounts of water vapor in the vacuum pump’s inlet vapor stream. This “water vapor tolerance” is the maximum inlet pressure point at which the pump is able to operate with pure water/fluids and at the same time prevents condensation inside the pump. Due to the high pumping speed and easy-to-clean, compact housing, this pump is ideally suited for use in laboratory applications within rough and fine vacuum ranges from 1 to 2 x 10–3 mbar.
1 Year Warranty
Specifications
  • DescriptionReplacment Vacuum Pump Oil