IKA Vacstar Digital Membrane Vacuum Pump; 1/Ea
Representative image only.
IKA Vacstar Digital Membrane Vacuum Pump; 1/Ea
IKA | Mfr: 20016241| Item: 1635460
Provides high suction capacity in a compact design
₹255,564.00INR / Each
₹255,564.00INR / Each
Product Description
Includes
USB cable, vacuum hose (1 m), connecting cable, silencer and mains cable
Key Features
  • Quiet operation—rated at or below 54 dB(A)
  • Membranes of the pump are resistant to chemicals
More About this Item
The VACSTAR digital dry four-chamber membrane vacuum pumps offer a powerful suction performance of 22 L/min at a minimum pressure of 2 mbar. The compact, portable pumps offer low-maintenance, are chemically resistant, and can be used for filtration, separation, and removal. Easily select your operating mode using the intuitive operator menu or the optional remote control. Set up all your parameters over a user-friendly display using a rotary push-button.
The pump enables you to construct your own laboratory vacuum system using any number of vacuum controllers. Automated processes can be controlled with the vacuum controller VC 10, which is available as an accessory. The pump is ideal for dry and oil-free applications in the laboratory.