IKA Vacstar Control Membrane Vacuum Pump; 1/Ea
Representative image only.
IKA Vacstar Control Membrane Vacuum Pump; 1/Ea
IKA | Mfr: 20019071| Item: 1635461
Provides high suction capacity in a compact design
₹377,840.00INR / Each
₹377,840.00INR / Each
Product Description
Includes
USB cable, USB micro cable, vacuum hose (1 m), vacuum safety emission condenser (VSE 1), vacuum hose (0.3 m) for VSE 1, wireless controller, screw driver for WiCo, cover, bracket, receiving flask, spherical clamp and mains cable
Key Features
  • Quiet operation—rated at or below 54 dB(A)
  • Membranes of the pump are resistant to chemicals
  • Vacuum safety emission condenser protects user and environment
More About this Item
The VACSTAR control dry four-chamber membrane vacuum pumps offer a powerful suction performance of 22 L/min at a minimum pressure of 2 mbar. The compact, portable pump offer low- maintenance, is chemically resistant, and can be used for filtration, separation, and removal. Easily select your operating mode using the intuitive operator menu or the optional remote control. Set up all your parameters over a user-friendly display using a rotary push-button.
The pump features a removable display and integrated vacuum wireless controller (WiCo) that can operate at a distance up to 150 m from the station using Bluetooth® connection. The controller offers a suitable mode for the desired process: from automatic to manual operation, right through to pumping or an evacuation and cleaning function. A downstream vacuum safety emission condenser (VSE1) protects you and the environment from harmful exhaust air.